Assay Details
| Gene Symbol: prkab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328554.1 | NP_001315483.1 | 3 - 4 | 424 | 76 | |
| NM_214713.2 | NP_999878.1 | 3 - 4 | 469 | 76 | ||
| GenBank mRNA | BC066758.1 | - | 3 - 4 | 384 | 76 | |
| EH456078.1 | - | 3 - 4 | 417 | 76 | ||
More Information
Set Membership: |
|

prkab1b
Genomic Map