Assay Details
| Gene Symbol: prkab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328554.1 | NP_001315483.1 | 7 - 8 | 838 | 68 | |
| NM_214713.2 | NP_999878.1 | 7 - 8 | 883 | 68 | ||
| GenBank mRNA | BC066758.1 | - | 7 - 8 | 798 | 68 | |
| EH456078.1 | - | 7 - 8 | 831 | 68 | ||
| EH461046.1 | - | 1 - 2 | 561 | 68 | ||
More Information
Set Membership: |
|

prkab1b
Genomic Map