Assay Details
| Gene Symbol: ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001951.1 | NP_001001951.1 | 9 - 10 | 1289 | 66 | |
| GenBank mRNA | BC063998.1 | - | 9 - 10 | 1282 | 66 | |
| BC074046.1 | - | 9 - 10 | 1289 | 66 | ||
More Information
Set Membership: |
|

ubxn7
Genomic Map