Assay Details
| Gene Symbol: cdk5rap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002105.1 | NP_001002105.1 | 12 - 13 | 1382 | 63 | |
| GenBank mRNA | BC071504.1 | - | 12 - 13 | 1382 | 63 | |
More Information
Set Membership: |
|

cdk5rap3
Genomic Map