Assay Details
| Gene Symbol: tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002208.1 | NP_001002208.1 | 4 - 5 | 468 | 70 | |
| XM_009295188.3 | XP_009293463.1 | 4 - 5 | 508 | 70 | ||
| GenBank mRNA | BC074075.1 | - | 4 - 5 | 468 | 70 | |
More Information
Set Membership: |
|

tpgs2
Genomic Map