Assay Details
| Gene Symbol: ip6k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002389.1 | NP_001002389.2 | 1 - 2 | 395 | 64 | |
| GenBank mRNA | BC076496.1 | - | 1 - 2 | 395 | 64 | |
More Information
Set Membership: |
|

ip6k2b
Genomic Map