Assay Details
| Gene Symbol: acp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002452.1 | NP_001002452.1 | 3 - 4 | 538 | 78 | |
| GenBank mRNA | BC076019.1 | - | 3 - 4 | 538 | 78 | |
| HM239641.1 | - | 3 - 4 | 370 | 78 | ||
| HM239642.1 | - | 3 - 4 | 369 | 78 | ||
More Information
Set Membership: |
|

acp5b
Genomic Map