Assay Details
| Gene Symbol: tspan33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002617.2 | NP_001002617.2 | 6 - 7 | 762 | 74 | |
| NM_001328582.1 | NP_001315511.1 | 5 - 6 | 707 | 74 | ||
| GenBank mRNA | BC075972.1 | - | 5 - 6 | 657 | 74 | |
| EH450479.1 | - | 6 - 7 | 789 | 74 | ||
More Information
Set Membership: |
|

tspan33a
Genomic Map