Assay Details
| Gene Symbol: slc2a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002643.1 | NP_001002643.1 | 3 - 4 | 373 | 64 | |
| XM_005159425.4 | XP_005159482.1 | 4 - 5 | 1259 | 64 | ||
| XM_005159426.4 | XP_005159483.1 | 4 - 5 | 758 | 64 | ||
| GenBank mRNA | BC076560.1 | - | 3 - 4 | 373 | 64 | |
More Information
Set Membership: |
|

slc2a3a
Genomic Map