Assay Details
| Gene Symbol: uqcrc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002657.2 | NP_001002657.2 | 13 - 14 | 1373 | 81 | |
| GenBank mRNA | BC076480.1 | - | 13 - 14 | 1342 | 81 | |
| BC164681.1 | - | 13 - 14 | 1333 | 81 | ||
More Information
Set Membership: |
|

uqcrc2a
Genomic Map