Assay Details
| Gene Symbol: bcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002676.2 | NP_001002676.2 | 6 - 7 | 1056 | 62 | |
| NM_001309527.1 | NP_001296456.1 | 6 - 7 | 888 | 62 | ||
| GenBank mRNA | BC076468.1 | - | 5 - 6 | 873 | 62 | |
| BC165882.1 | - | 4 - 5 | 797 | 62 | ||
More Information
Set Membership: |
|

bcat2
Genomic Map