Assay Details
| Gene Symbol: vps26c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003421.1 | NP_001003421.1 | 2 - 3 | 225 | 147 | |
| GenBank mRNA | BC075958.1 | - | 2 - 3 | 225 | 147 | |
More Information
Set Membership: |
|

vps26c
Genomic Map