Assay Details
| Gene Symbol: commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003553.2 | NP_001003553.1 | 3 - 4 | 325 | 97 | |
| GenBank mRNA | BC078284.1 | - | 3 - 4 | 330 | 97 | |
More Information
Set Membership: |
|

commd5
Genomic Map