Assay Details
| Gene Symbol: cibar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003604.2 | NP_001003604.1 | 6 - 7 | 637 | 132 | |
| GenBank mRNA | BC078261.1 | - | 6 - 7 | 641 | 132 | |
More Information
Set Membership: |
|

cibar1
Genomic Map