Assay Details
| Gene Symbol: fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003634.1 | NP_001003634.1 | 3 - 4 | 386 | 59 | |
| XM_005166828.4 | XP_005166885.1 | 3 - 4 | 382 | 59 | ||
| XM_021478241.1 | XP_021333916.1 | 3 - 4 | 382 | 59 | ||
| XM_021478242.1 | XP_021333917.1 | 3 - 4 | 383 | 59 | ||
| GenBank mRNA | BC077109.1 | - | 3 - 4 | 386 | 59 | |
More Information
Set Membership: |
|

fnbp1l
Genomic Map