Assay Details
| Gene Symbol: babam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003752.2 | NP_001003752.2 | 5 - 6 | 756 | 60 | |
| XM_017358217.2 | XP_017213706.1 | 4 - 5 | 717 | 60 | ||
| GenBank mRNA | BC079500.1 | - | 5 - 6 | 762 | 60 | |
| CN323673.1 | - | 3 - 4 | 382 | 60 | ||
| EH490619.1 | - | 5 - 6 | 762 | 60 | ||
| EH516962.1 | - | 5 - 6 | 541 | 60 | ||
More Information
Set Membership: |
|

babam1
Genomic Map