Assay Details
| Gene Symbol: coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003823.1 | NP_001003823.1 | 3 - 4 | 149 | 68 | |
| GenBank mRNA | BC162181.1 | - | 3 - 4 | 123 | 68 | |
| BC162194.1 | - | 3 - 4 | 123 | 68 | ||
More Information
Set Membership: |
|

coch
Genomic Map