Assay Details
| Gene Symbol: sdhdb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004004.2 | NP_001004004.1 | 3 - 4 | 335 | 76 | |
| GenBank mRNA | BC079507.1 | - | 3 - 4 | 337 | 76 | |
More Information
Set Membership: |
|

sdhdb
Genomic Map