Assay Details
| Gene Symbol: emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004119.1 | NP_001004119.1 | 4 - 5 | 476 | 61 | |
| XM_021470381.1 | XP_021326056.1 | 4 - 5 | 502 | 61 | ||
| GenBank mRNA | BC080211.1 | - | 4 - 5 | 476 | 61 | |
More Information
Set Membership: |
|

emp2
Genomic Map