Assay Details
| Gene Symbol: lyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004543.1 | NP_001004543.1 | 8 - 9 | 928 | 62 | |
| XM_005163399.4 | XP_005163456.1 | 8 - 9 | 883 | 62 | ||
| GenBank mRNA | BC081601.1 | - | 8 - 9 | 928 | 62 | |
More Information
Set Membership: |
|

lyn
Genomic Map