Assay Details
| Gene Symbol: rimkla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004554.2 | NP_001004554.1 | 4 - 5 | 1109 | 57 | |
| XM_009303991.3 | XP_009302266.1 | 5 - 6 | 710 | 57 | ||
| GenBank mRNA | BC081615.1 | - | 4 - 5 | 1109 | 57 | |
More Information
Set Membership: |
|

rimkla
Genomic Map