Assay Details
| Gene Symbol: slc25a24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004606.1 | NP_001004606.1 | 9 - 10 | 1297 | 58 | |
| XM_005160680.4 | XP_005160737.1 | 6 - 7 | 1021 | 58 | ||
| GenBank mRNA | BC078435.1 | - | 9 - 10 | 1297 | 58 | |
More Information
Set Membership: |
|

slc25a24
Genomic Map