Assay Details
| Gene Symbol: slc66a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004615.1 | NP_001004615.1 | 4 - 5 | 475 | 77 | |
| GenBank mRNA | BC081478.1 | - | 4 - 5 | 475 | 77 | |
More Information
Set Membership: |
|

slc66a3
Genomic Map