Assay Details
| Gene Symbol: ctsz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006043.1 | NP_001006043.1 | 4 - 5 | 629 | 87 | |
| GenBank mRNA | BC083369.1 | - | 4 - 5 | 629 | 87 | |
| BC164067.1 | - | 4 - 5 | 641 | 87 | ||
More Information
Set Membership: |
|

ctsz
Genomic Map