Assay Details
| Gene Symbol: gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006103.2 | NP_001006103.2 | 6 - 7 | 1033 | 58 | |
| GenBank mRNA | BC083209.1 | - | 6 - 7 | 1034 | 58 | |
| BC164755.1 | - | 5 - 6 | 782 | 58 | ||
More Information
Set Membership: |
|

gla
Genomic Map