Assay Details
| Gene Symbol: trpa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007065.1 | NP_001007066.1 | 12 - 13 | 1383 | 83 | |
| XM_017358267.2 | XP_017213756.1 | 11 - 12 | 1378 | 83 | ||
| GenBank mRNA | AY677196.1 | - | 12 - 13 | 1383 | 83 | |
| EU826643.1 | - | 11 - 12 | 1352 | 83 | ||
More Information
Set Membership: |
|

trpa1a
Genomic Map