Assay Details
| Gene Symbol: trpa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007065.1 | NP_001007066.1 | 9 - 10 | 1014 | 103 | |
| XM_017358267.2 | XP_017213756.1 | 8 - 9 | 1009 | 103 | ||
| GenBank mRNA | AY677196.1 | - | 9 - 10 | 1014 | 103 | |
| EU826643.1 | - | 8 - 9 | 983 | 103 | ||
More Information
Set Membership: |
|

trpa1a
Genomic Map