Assay Details
| Gene Symbol: pptc7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007378.1 | NP_001007379.1 | 1 - 2 | 515 | 102 | |
| GenBank mRNA | BC085459.1 | - | 1 - 2 | 515 | 102 | |
More Information
Set Membership: |
|

pptc7a
Genomic Map