Assay Details
| Gene Symbol: clk4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013294.3 | NP_001013312.2 | 6 - 7 | 972 | 86 | |
| NM_001329908.1 | NP_001316837.1 | 10 - 11 | 1226 | 86 | ||
| XM_009295587.3 | XP_009293862.1 | 7 - 8 | 789 | 86 | ||
| GenBank mRNA | BC090270.1 | - | 6 - 7 | 899 | 86 | |
| BC164422.1 | - | 4 - 5 | 493 | 86 | ||
| CT646097.2 | - | 3 - 4 | 512 | 86 | ||
| EH458583.1 | - | 3 - 4 | 494 | 86 | ||
More Information
Set Membership: |
|

clk4b
Genomic Map