Assay Details
| Gene Symbol: cbsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014345.2 | NP_001014367.2 | 11 - 12 | 1397 | 63 | |
| GenBank mRNA | AY880251.1 | - | 11 - 12 | 1397 | 63 | |
| BC091981.1 | - | 7 - 8 | 738 | 63 | ||
| BC164709.1 | - | 6 - 7 | 734 | 63 | ||
More Information
Set Membership: |
|

cbsb
Genomic Map