Assay Details
| Gene Symbol: cbsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014345.2 | NP_001014367.2 | 4 - 5 | 622 | 90 | |
| GenBank mRNA | AY880251.1 | - | 4 - 5 | 622 | 90 | |
More Information
Set Membership: |
|

cbsb
Genomic Map