Assay Details
| Gene Symbol: cbsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014345.2 | NP_001014367.2 | 7 - 8 | 1005 | 81 | |
| GenBank mRNA | AY880251.1 | - | 7 - 8 | 1005 | 81 | |
| BC091981.1 | - | 3 - 4 | 346 | 81 | ||
| BC164709.1 | - | 2 - 3 | 342 | 81 | ||
More Information
Set Membership: |
|

cbsb
Genomic Map