Assay Details
| Gene Symbol: erbb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014826.1 | NP_001014826.1 | 15 - 16 | 2161 | 79 | |
| XM_005162170.4 | XP_005162227.1 | 16 - 17 | 2261 | 79 | ||
| XM_021469802.1 | XP_021325477.1 | 15 - 16 | 2253 | 79 | ||
| GenBank mRNA | AY863052.1 | - | 15 - 16 | 2161 | 79 | |
| BC162634.1 | - | 16 - 17 | 2051 | 79 | ||
| Gene Symbol: LOC100149258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128819.1 | NP_001122291.1 | 16 - 17 | 2051 | 79 | |
More Information
Set Membership: |
|

erbb3b
Genomic Map