Assay Details
| Gene Symbol: erbb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014826.1 | NP_001014826.1 | 24 - 25 | 3239 | 148 | |
| XM_021469802.1 | XP_021325477.1 | 24 - 25 | 3331 | 148 | ||
| GenBank mRNA | AY863052.1 | - | 24 - 25 | 3239 | 148 | |
More Information
Set Membership: |
|

erbb3b
Genomic Map