Assay Details
| Gene Symbol: erbb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014826.1 | NP_001014826.1 | 27 - 28 | 3737 | 109 | |
| XM_005162170.4 | XP_005162227.1 | 28 - 29 | 3831 | 109 | ||
| XM_021469802.1 | XP_021325477.1 | 27 - 28 | 3829 | 109 | ||
| GenBank mRNA | AY863052.1 | - | 27 - 28 | 3737 | 109 | |
| BC162634.1 | - | 28 - 29 | 3627 | 109 | ||
| Gene Symbol: LOC100149258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128819.1 | NP_001122291.1 | 28 - 29 | 3627 | 109 | |
More Information
Set Membership: |
|

erbb3b
Genomic Map