Assay Details
| Gene Symbol: marcksb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015060.2 | NP_001015060.2 | 1 - 2 | 330 | 85 | |
| GenBank mRNA | BC055217.1 | - | 1 - 2 | 101 | 85 | |
| BC092814.1 | - | 1 - 2 | 281 | 85 | ||
| BC165144.1 | - | 1 - 2 | 120 | 85 | ||
| EH503964.1 | - | 1 - 2 | 364 | 85 | ||
| GU563329.1 | - | 1 - 2 | 86 | 85 | ||
| JF894243.1 | - | 1 - 2 | 152 | 85 | ||
More Information
Set Membership: |
|

marcksb
Genomic Map