Assay Details
| Gene Symbol: rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017358.1 | NP_001017358.1 | 3 - 4 | 1011 | 92 | |
| GenBank mRNA | AB035930.1 | - | 3 - 4 | 1011 | 92 | |
| BC116527.1 | - | 3 - 4 | 812 | 92 | ||
| BC121755.1 | - | 3 - 4 | 1028 | 92 | ||
| BC139514.1 | - | 3 - 4 | 824 | 92 | ||
More Information
Set Membership: |
|

rspo3
Genomic Map