Assay Details
| Gene Symbol: phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017823.1 | NP_001017823.1 | 6 - 7 | 755 | 62 | |
| GenBank mRNA | BC092793.1 | - | 6 - 7 | 755 | 62 | |
| BC164367.1 | - | 6 - 7 | 704 | 62 | ||
More Information
Set Membership: |
|

phyh
Genomic Map