Assay Details
| Gene Symbol: arl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017827.2 | NP_001017827.1 | 6 - 7 | 554 | 101 | |
| NM_001328421.1 | NP_001315350.1 | 5 - 6 | 557 | 101 | ||
| XM_009307160.3 | XP_009305435.1 | 5 - 6 | 550 | 101 | ||
| GenBank mRNA | BC092789.1 | - | 6 - 7 | 101 | ||
More Information
Set Membership: |
|

arl3a
Genomic Map