Assay Details
| Gene Symbol: vkorc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020688.1 | NP_001018524.1 | 1 - 2 | 213 | 82 | |
| GenBank mRNA | BC095778.1 | - | 1 - 2 | 213 | 82 | |
| BC164023.1 | - | 1 - 2 | 231 | 82 | ||
More Information
Set Membership: |
|

vkorc1l1
Genomic Map