Assay Details
| Gene Symbol: sept3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024418.2 | NP_001019589.2 | 5 - 6 | 787 | 85 | |
| XM_005160040.4 | XP_005160097.1 | 5 - 6 | 701 | 85 | ||
| GenBank mRNA | BC095158.1 | - | 5 - 6 | 787 | 85 | |
More Information
Set Membership: |
|

sept3
Genomic Map