Assay Details
| Gene Symbol: lta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024821.1 | NP_001019992.1 | 4 - 5 | 243 | 114 | |
| XM_009291801.3 | XP_009290076.1 | 5 - 6 | 397 | 114 | ||
| GenBank mRNA | AB183468.1 | - | 4 - 5 | 243 | 114 | |
More Information
Set Membership: |
|

lta
Genomic Map