Assay Details
| Gene Symbol: stxbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025182.1 | NP_001020353.1 | 13 - 14 | 1140 | 71 | |
| XM_005161113.4 | XP_005161170.1 | 13 - 14 | 1215 | 71 | ||
| GenBank mRNA | BC097221.1 | - | 13 - 14 | 1140 | 71 | |
More Information
Set Membership: |
|

stxbp1a
Genomic Map