Assay Details
| Gene Symbol: stxbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025182.1 | NP_001020353.1 | 18 - 19 | 1730 | 58 | |
| GenBank mRNA | BC097221.1 | - | 18 - 19 | 1730 | 58 | |
More Information
Set Membership: |
|

stxbp1a
Genomic Map