Assay Details
| Gene Symbol: smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025535.2 | NP_001020706.2 | 1 - 2 | 268 | 89 | |
| XM_005173126.4 | XP_005173183.1 | 1 - 2 | 265 | 89 | ||
| GenBank mRNA | BC083218.1 | - | 1 - 2 | 162 | 89 | |
| BC097106.1 | - | 1 - 2 | 256 | 89 | ||
| EH457174.1 | - | 1 - 2 | 285 | 89 | ||
More Information
Set Membership: |
|

smim19
Genomic Map