Assay Details
| Gene Symbol: smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025535.2 | NP_001020706.2 | 2 - 3 | 392 | 81 | |
| GenBank mRNA | BC097106.1 | - | 2 - 3 | 380 | 81 | |
| EH457174.1 | - | 2 - 3 | 409 | 81 | ||
| EH482896.1 | - | 1 - 2 | 633 | 81 | ||
More Information
Set Membership: |
|

smim19
Genomic Map