Assay Details
| Gene Symbol: nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029968.1 | NP_001025139.1 | 4 - 5 | 971 | 60 | |
| GenBank mRNA | BC098537.1 | - | 4 - 5 | 971 | 60 | |
More Information
Set Membership: |
|

nif3l1
Genomic Map