Assay Details
| Gene Symbol: nabp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030252.1 | NP_001025423.1 | 4 - 5 | 618 | 85 | |
| GenBank mRNA | BC097147.1 | - | 4 - 5 | 618 | 85 | |
More Information
Set Membership: |
|

nabp1b
Genomic Map