Assay Details
| Gene Symbol: slc4a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034984.1 | NP_001030156.1 | 14 - 15 | 1924 | 119 | |
| XM_005165244.4 | XP_005165301.1 | 14 - 15 | 119 | |||
| XM_009301742.3 | XP_009300017.1 | 14 - 15 | 119 | |||
| XM_009301743.3 | XP_009300018.1 | 13 - 14 | 119 | |||
| XM_009301747.3 | XP_009300022.1 | 13 - 14 | 119 | |||
| XM_021476291.1 | XP_021331966.1 | 16 - 17 | 119 | |||
| XM_021476293.1 | XP_021331968.1 | 16 - 17 | 119 | |||
| XM_021476294.1 | XP_021331969.1 | 16 - 17 | 119 | |||
| GenBank mRNA | AY727858.1 | - | 14 - 15 | 1924 | 119 | |
More Information
Set Membership: |
|

slc4a4a
Genomic Map