Assay Details
| Gene Symbol: slc4a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034984.1 | NP_001030156.1 | 20 - 21 | 2711 | 86 | |
| XM_005165244.4 | XP_005165301.1 | 20 - 21 | 2961 | 86 | ||
| XM_009301742.3 | XP_009300017.1 | 20 - 21 | 3015 | 86 | ||
| XM_009301743.3 | XP_009300018.1 | 19 - 20 | 2918 | 86 | ||
| XM_009301747.3 | XP_009300022.1 | 19 - 20 | 2891 | 86 | ||
| XM_021476291.1 | XP_021331966.1 | 22 - 23 | 3004 | 86 | ||
| XM_021476293.1 | XP_021331968.1 | 22 - 23 | 2977 | 86 | ||
| XM_021476294.1 | XP_021331969.1 | 22 - 23 | 2946 | 86 | ||
| GenBank mRNA | AY727858.1 | - | 20 - 21 | 2711 | 86 | |
| BC162559.1 | - | 20 - 21 | 2743 | 86 | ||
More Information
Set Membership: |
|

slc4a4a
Genomic Map